Learn More
Hampton Research HR2-998-23/PEG/Ion 2 Screen #23 (F11), 185 ml - 0.1 M DL-Malic acid pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502475
Encompass
HR2-998-23/PEG/Ion 2 Screen #23 (F11), 185 ml - 0.1 M DL-Malic acid pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-5 Supplier Hampton Research Supplier No. HR299823
HR2-998-23/PEG/Ion 2 Screen #23 (F11), 185 ml - 0.1 M DL-Malic acid pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.