Learn More
Hampton Research HR2-998-25/PEG/Ion 2 Screen #25 (G01), 185 ml - 0.1 M Sodium acetate trihydrate pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502477
Encompass
HR2-998-25/PEG/Ion 2 Screen #25 (G01), 185 ml - 0.1 M Sodium acetate trihydrate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-7 Supplier Hampton Research Supplier No. HR299825
HR2-998-25/PEG/Ion 2 Screen #25 (G01), 185 ml - 0.1 M Sodium acetate trihydrate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.