Learn More
Hampton Research HR2-998-26/PEG/Ion 2 Screen #26 (G02), 185 ml - 0.2 M Sodium acetate trihydrate pH 7.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502478
Encompass
HR2-998-26/PEG/Ion 2 Screen #26 (G02), 185 ml - 0.2 M Sodium acetate trihydrate pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-8 Supplier Hampton Research Supplier No. HR299826
HR2-998-26/PEG/Ion 2 Screen #26 (G02), 185 ml - 0.2 M Sodium acetate trihydrate pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.