Learn More
Hampton Research HR2-998-27/PEG/Ion 2 Screen #27 (G03), 185 ml - 0.1 M Sodium formate pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502479
Encompass
HR2-998-27/PEG/Ion 2 Screen #27 (G03), 185 ml - 0.1 M Sodium formate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-9 Supplier Hampton Research Supplier No. HR299827
HR2-998-27/PEG/Ion 2 Screen #27 (G03), 185 ml - 0.1 M Sodium formate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.