Learn More
Hampton Research HR2-998-28/PEG/Ion 2 Screen #28 (G04), 185 ml - 0.2 M Sodium formate pH 7.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502480
Encompass
HR2-998-28/PEG/Ion 2 Screen #28 (G04), 185 ml - 0.2 M Sodium formate pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-0 Supplier Hampton Research Supplier No. HR299828
HR2-998-28/PEG/Ion 2 Screen #28 (G04), 185 ml - 0.2 M Sodium formate pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.