Learn More
Hampton Research HR2-998-29/PEG/Ion 2 Screen #29 (G05), 185 ml - 0.1 M Ammonium tartrate dibasic pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502481
Encompass
HR2-998-29/PEG/Ion 2 Screen #29 (G05), 185 ml - 0.1 M Ammonium tartrate dibasic pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-1 Supplier Hampton Research Supplier No. HR299829
HR2-998-29/PEG/Ion 2 Screen #29 (G05), 185 ml - 0.1 M Ammonium tartrate dibasic pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.