Learn More
Hampton Research HR2-998-30/PEG/Ion 2 Screen #30 (G06), 185 ml - 0.2 M Ammonium tartrate dibasic pH 7.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502482
Encompass
HR2-998-30/PEG/Ion 2 Screen #30 (G06), 185 ml - 0.2 M Ammonium tartrate dibasic pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-2 Supplier Hampton Research Supplier No. HR299830
HR2-998-30/PEG/Ion 2 Screen #30 (G06), 185 ml - 0.2 M Ammonium tartrate dibasic pH 7.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.