Learn More
Hampton Research HR2-998-31/PEG/Ion 2 Screen #31 (G07), 185 ml - 2% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.6, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502483
Encompass
HR2-998-31/PEG/Ion 2 Screen #31 (G07), 185 ml - 2% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.6, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-3 Supplier Hampton Research Supplier No. HR299831
HR2-998-31/PEG/Ion 2 Screen #31 (G07), 185 ml - 2% v/v Tacsimate pH 4.0, 0.1 M Sodium acetate trihydrate pH 4.6, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.