Learn More
Hampton Research HR2-998-32/PEG/Ion 2 Screen #32 (G08), 185 ml - 2% v/v Tacsimate pH 5.0, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502484
Encompass
HR2-998-32/PEG/Ion 2 Screen #32 (G08), 185 ml - 2% v/v Tacsimate pH 5.0, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-4 Supplier Hampton Research Supplier No. HR299832
HR2-998-32/PEG/Ion 2 Screen #32 (G08), 185 ml - 2% v/v Tacsimate pH 5.0, 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.