Learn More
Hampton Research HR2-998-34/PEG/Ion 2 Screen #34 (G10), 185 ml - 2% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502486
Encompass
HR2-998-34/PEG/Ion 2 Screen #34 (G10), 185 ml - 2% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-6 Supplier Hampton Research Supplier No. HR299834
HR2-998-34/PEG/Ion 2 Screen #34 (G10), 185 ml - 2% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.5, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.