Learn More
Hampton Research HR2-998-35/PEG/Ion 2 Screen #35 (G11), 185 ml - 2% v/v Tacsimate pH 8.0, 0.1 M Tris pH 8.5, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502487
Encompass
HR2-998-35/PEG/Ion 2 Screen #35 (G11), 185 ml - 2% v/v Tacsimate pH 8.0, 0.1 M Tris pH 8.5, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-7 Supplier Hampton Research Supplier No. HR299835
HR2-998-35/PEG/Ion 2 Screen #35 (G11), 185 ml - 2% v/v Tacsimate pH 8.0, 0.1 M Tris pH 8.5, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.