Learn More
Hampton Research HR2-998-36/PEG/Ion 2 Screen #36 (G12), 185 ml - (0.07 M Citric acid, 0.03 M BIS-TRIS propane)/pH 3.4, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502488
Encompass
HR2-998-36/PEG/Ion 2 Screen #36 (G12), 185 ml - (0.07 M Citric acid, 0.03 M BIS-TRIS propane)/pH 3.4, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-8 Supplier Hampton Research Supplier No. HR299836
HR2-998-36/PEG/Ion 2 Screen #36 (G12), 185 ml - (0.07 M Citric acid, 0.03 M BIS-TRIS propane)/pH 3.4, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.