Learn More
Hampton Research HR2-998-37/PEG/Ion 2 Screen #37 (H01), 185 ml - (0.06 M Citric acid, 0.04 M BIS-TRIS propane)/pH 4.1, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502489
Encompass
HR2-998-37/PEG/Ion 2 Screen #37 (H01), 185 ml - (0.06 M Citric acid, 0.04 M BIS-TRIS propane)/pH 4.1, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-248-9 Supplier Hampton Research Supplier No. HR299837
HR2-998-37/PEG/Ion 2 Screen #37 (H01), 185 ml - (0.06 M Citric acid, 0.04 M BIS-TRIS propane)/pH 4.1, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.