Learn More
Hampton Research HR2-998-38/PEG/Ion 2 Screen #38 (H02), 185 ml - (0.05 M Citric acid, 0.05 M BIS-TRIS propane)/pH 5.0, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502490
Encompass
HR2-998-38/PEG/Ion 2 Screen #38 (H02), 185 ml - (0.05 M Citric acid, 0.05 M BIS-TRIS propane)/pH 5.0, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-0 Supplier Hampton Research Supplier No. HR299838
HR2-998-38/PEG/Ion 2 Screen #38 (H02), 185 ml - (0.05 M Citric acid, 0.05 M BIS-TRIS propane)/pH 5.0, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.