Learn More
Hampton Research HR2-998-39/PEG/Ion 2 Screen #39 (H03), 185 ml - (0.04 M Citric acid, 0.06 M BIS-TRIS propane)/pH 6.4, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502491
Encompass
HR2-998-39/PEG/Ion 2 Screen #39 (H03), 185 ml - (0.04 M Citric acid, 0.06 M BIS-TRIS propane)/pH 6.4, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-1 Supplier Hampton Research Supplier No. HR299839
HR2-998-39/PEG/Ion 2 Screen #39 (H03), 185 ml - (0.04 M Citric acid, 0.06 M BIS-TRIS propane)/pH 6.4, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.