Learn More
Hampton Research HR2-998-41/PEG/Ion 2 Screen #41 (H05), 185 ml - (0.02 M Citric acid, 0.08 M BIS-TRIS propane)/pH 8.8, 16% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502493
Encompass
HR2-998-41/PEG/Ion 2 Screen #41 (H05), 185 ml - (0.02 M Citric acid, 0.08 M BIS-TRIS propane)/pH 8.8, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-3 Supplier Hampton Research Supplier No. HR299841
HR2-998-41/PEG/Ion 2 Screen #41 (H05), 185 ml - (0.02 M Citric acid, 0.08 M BIS-TRIS propane)/pH 8.8, 16% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.