Learn More
Hampton Research HR2-998-43/PEG/Ion 2 Screen #43 (H07), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.005 M Nickel(II) chloride hexahydrate, 0.1 M HEPES sodium pH 7.0, 15% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502495
Encompass
HR2-998-43/PEG/Ion 2 Screen #43 (H07), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.005 M Nickel(II) chloride hexahydrate, 0.1 M HEPES sodium pH 7.0, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-5 Supplier Hampton Research Supplier No. HR299843
HR2-998-43/PEG/Ion 2 Screen #43 (H07), 185 ml - 0.01 M Magnesium chloride hexahydrate, 0.005 M Nickel(II) chloride hexahydrate, 0.1 M HEPES sodium pH 7.0, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.