Learn More
Hampton Research HR2-998-45/PEG/Ion 2 Screen #45 (H09), 185 ml - 0.15 M Cesium chloride, 15% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502497
Encompass
HR2-998-45/PEG/Ion 2 Screen #45 (H09), 185 ml - 0.15 M Cesium chloride, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-7 Supplier Hampton Research Supplier No. HR299845
HR2-998-45/PEG/Ion 2 Screen #45 (H09), 185 ml - 0.15 M Cesium chloride, 15% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.