Learn More
Hampton Research HR2-998-46/PEG/Ion 2 Screen #46 (H10), 185 ml - 0.2 M Sodium bromide, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502498
Encompass
HR2-998-46/PEG/Ion 2 Screen #46 (H10), 185 ml - 0.2 M Sodium bromide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-249-8 Supplier Hampton Research Supplier No. HR299846
HR2-998-46/PEG/Ion 2 Screen #46 (H10), 185 ml - 0.2 M Sodium bromide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.