Learn More
Hampton Research PEG/ION SCREEN NO01 185ML
SDP
Catalog No. 501824
Encompass
HR2-922-01/PEG/Ion Screen #01 (A1), 185 ml - 0.2 M Sodium fluoride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-182-4 Supplier Hampton Research Supplier No. HR292201
HR2-922-01/PEG/Ion Screen #01 (A1), 185 ml - 0.2 M Sodium fluoride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.