Learn More
Hampton Research PEG/ION SCREEN NO02 185ML
SDP
Catalog No. 501825
Encompass
HR2-922-02/PEG/Ion Screen #02 (A2), 185 ml - 0.2 M Potassium fluoride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-182-5 Supplier Hampton Research Supplier No. HR292202
HR2-922-02/PEG/Ion Screen #02 (A2), 185 ml - 0.2 M Potassium fluoride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.