Learn More
Hampton Research PEG/ION SCREEN NO04 185ML
SDP
Catalog No. 501827
Encompass
HR2-922-04/PEG/Ion Screen #04 (A4), 185 ml - 0.2 M Lithium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-182-7 Supplier Hampton Research Supplier No. HR292204
HR2-922-04/PEG/Ion Screen #04 (A4), 185 ml - 0.2 M Lithium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.