Learn More
Hampton Research PEG/ION SCREEN NO07 185ML
SDP
Catalog No. 501830
Encompass
HR2-922-07/PEG/Ion Screen #07 (A7), 185 ml - 0.2 M Calcium chloride dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-0 Supplier Hampton Research Supplier No. HR292207
HR2-922-07/PEG/Ion Screen #07 (A7), 185 ml - 0.2 M Calcium chloride dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.