Learn More
Hampton Research PEG/ION SCREEN NO08 185ML
SDP
Catalog No. 501831
Encompass
HR2-922-08/PEG/Ion Screen #08 (A8), 185 ml - 0.2 M Potassium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-1 Supplier Hampton Research Supplier No. HR292208
HR2-922-08/PEG/Ion Screen #08 (A8), 185 ml - 0.2 M Potassium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.