Learn More
Hampton Research PEG/ION SCREEN NO09 185ML
SDP
Catalog No. 501832
Encompass
HR2-922-09/PEG/Ion Screen #09 (A9), 185 ml - 0.2 M Ammonium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-2 Supplier Hampton Research Supplier No. HR292209
HR2-922-09/PEG/Ion Screen #09 (A9), 185 ml - 0.2 M Ammonium chloride, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.