Learn More
Hampton Research PEG/ION SCREEN NO10 185ML
SDP
Catalog No. 501833
Encompass
HR2-922-10/PEG/Ion Screen #10 (A10), 185 ml - 0.2 M Sodium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-3 Supplier Hampton Research Supplier No. HR292210
HR2-922-10/PEG/Ion Screen #10 (A10), 185 ml - 0.2 M Sodium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.