Learn More
Hampton Research PEG/ION SCREEN NO11 185ML
SDP
Catalog No. 501834
Encompass
HR2-922-11/PEG/Ion Screen #11 (A11), 185 ml - 0.2 M Potassium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-4 Supplier Hampton Research Supplier No. HR292211
HR2-922-11/PEG/Ion Screen #11 (A11), 185 ml - 0.2 M Potassium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.