Learn More
Hampton Research PEG/ION SCREEN NO12 185ML
SDP
Catalog No. 501835
Encompass
HR2-922-12/PEG/Ion Screen #12 (A12), 185 ml - 0.2 M Ammonium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-5 Supplier Hampton Research Supplier No. HR292212
HR2-922-12/PEG/Ion Screen #12 (A12), 185 ml - 0.2 M Ammonium iodide, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.