Learn More
Hampton Research PEG/ION SCREEN NO13 185ML
SDP
Catalog No. 501836
Encompass
HR2-922-13/PEG/Ion Screen #13 (B1), 185 ml - 0.2 M Sodium thiocyanate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-6 Supplier Hampton Research Supplier No. HR292213
HR2-922-13/PEG/Ion Screen #13 (B1), 185 ml - 0.2 M Sodium thiocyanate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.