Learn More
Hampton Research PEG/ION SCREEN NO14 185ML
SDP
Catalog No. 501837
Encompass
HR2-922-14/PEG/Ion Screen #14 (B2), 185 ml - 0.2 M Potassium thiocyanate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-7 Supplier Hampton Research Supplier No. HR292214
HR2-922-14/PEG/Ion Screen #14 (B2), 185 ml - 0.2 M Potassium thiocyanate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.