Learn More
Hampton Research PEG/ION SCREEN NO16 185ML
SDP
Catalog No. 501839
Encompass
HR2-922-16/PEG/Ion Screen #16 (B4), 185 ml - 0.2 M Magnesium nitrate hexahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-183-9 Supplier Hampton Research Supplier No. HR292216
HR2-922-16/PEG/Ion Screen #16 (B4), 185 ml - 0.2 M Magnesium nitrate hexahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.