Learn More
Hampton Research PEG/ION SCREEN NO17 185ML
SDP
Catalog No. 501840
Encompass
HR2-922-17/PEG/Ion Screen #17 (B5), 185 ml - 0.2 M Sodium nitrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-0 Supplier Hampton Research Supplier No. HR292217
HR2-922-17/PEG/Ion Screen #17 (B5), 185 ml - 0.2 M Sodium nitrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.