Learn More
Hampton Research PEG/ION SCREEN NO18 185ML
SDP
Supplier:  Hampton Research HR292218
Encompass
HR2-922-18/PEG/Ion Screen #18 (B6), 185 ml - 0.2 M Potassium nitrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-1