Learn More
Hampton Research PEG/ION SCREEN NO19 185ML
SDP
Catalog No. 501842
Encompass
HR2-922-19/PEG/Ion Screen #19 (B7), 185 ml - 0.2 M Ammonium nitrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-2 Supplier Hampton Research Supplier No. HR292219
HR2-922-19/PEG/Ion Screen #19 (B7), 185 ml - 0.2 M Ammonium nitrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.