Learn More
Hampton Research PEG/ION SCREEN NO20 185ML
SDP
Catalog No. 501843
Encompass
HR2-922-20/PEG/Ion Screen #20 (B8), 185 ml - 0.2 M Magnesium formate dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-3 Supplier Hampton Research Supplier No. HR292220
HR2-922-20/PEG/Ion Screen #20 (B8), 185 ml - 0.2 M Magnesium formate dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.