Learn More
Hampton Research PEG/ION SCREEN NO22 185ML
SDP
Catalog No. 501845
Encompass
HR2-922-22/PEG/Ion Screen #22 (B10), 185 ml - 0.2 M Potassium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-5 Supplier Hampton Research Supplier No. HR292222
HR2-922-22/PEG/Ion Screen #22 (B10), 185 ml - 0.2 M Potassium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.