Learn More
Hampton Research PEG/ION SCREEN NO23 185ML
SDP
Catalog No. 501846
Encompass
HR2-922-23/PEG/Ion Screen #23 (B11), 185 ml - 0.2 M Ammonium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-6 Supplier Hampton Research Supplier No. HR292223
HR2-922-23/PEG/Ion Screen #23 (B11), 185 ml - 0.2 M Ammonium formate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.