Learn More
Hampton Research PEG/ION SCREEN NO24 185ML
SDP
Catalog No. 501847
Encompass
HR2-922-24/PEG/Ion Screen #24 (B12), 185 ml - 0.2 M Lithium acetate dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-7 Supplier Hampton Research Supplier No. HR292224
HR2-922-24/PEG/Ion Screen #24 (B12), 185 ml - 0.2 M Lithium acetate dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.