Learn More
Hampton Research PEG/ION SCREEN NO25 185ML
SDP
Catalog No. 501848
Encompass
HR2-922-25/PEG/Ion Screen #25 (C1), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-184-8 Supplier Hampton Research Supplier No. HR292225
HR2-922-25/PEG/Ion Screen #25 (C1), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.