Learn More
Hampton Research PEG/ION SCREEN NO27 185ML
SDP
Catalog No. 501850
Encompass
HR2-922-27/PEG/Ion Screen #27 (C3), 185 ml - 0.2 M Sodium acetate trihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-0 Supplier Hampton Research Supplier No. HR292227
HR2-922-27/PEG/Ion Screen #27 (C3), 185 ml - 0.2 M Sodium acetate trihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.