Learn More
Hampton Research PEG/ION SCREEN NO29 185ML
SDP
Catalog No. 501852
Encompass
HR2-922-29/PEG/Ion Screen #29 (C5), 185 ml - 0.2 M Potassium acetate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-2 Supplier Hampton Research Supplier No. HR292229
HR2-922-29/PEG/Ion Screen #29 (C5), 185 ml - 0.2 M Potassium acetate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.