Learn More
Hampton Research PEG/ION SCREEN NO33 185ML
SDP
Catalog No. 501856
Encompass
HR2-922-33/PEG/Ion Screen #33 (C9), 185 ml - 0.2 M Sodium sulfate decahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-6 Supplier Hampton Research Supplier No. HR292233
HR2-922-33/PEG/Ion Screen #33 (C9), 185 ml - 0.2 M Sodium sulfate decahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.