Learn More
Hampton Research PEG/ION SCREEN NO35 185ML
SDP
Catalog No. 501858
Encompass
HR2-922-35/PEG/Ion Screen #35 (C11), 185 ml - 0.2 M Ammonium sulfate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-8 Supplier Hampton Research Supplier No. HR292235
HR2-922-35/PEG/Ion Screen #35 (C11), 185 ml - 0.2 M Ammonium sulfate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.