Learn More
Hampton Research PEG/ION SCREEN NO36 185ML
SDP
Catalog No. 501859
Encompass
HR2-922-36/PEG/Ion Screen #36 (C12), 185 ml - 0.2 M Sodium tartrate dibasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-185-9 Supplier Hampton Research Supplier No. HR292236
HR2-922-36/PEG/Ion Screen #36 (C12), 185 ml - 0.2 M Sodium tartrate dibasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.