Learn More
Hampton Research PEG/ION SCREEN NO37 185ML
SDP
Supplier:  Hampton Research HR292237
Encompass
HR2-922-37/PEG/Ion Screen #37 (D1), 185 ml - 0.2 M Potassium sodium tartrate tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-0