Learn More
Hampton Research PEG/ION SCREEN NO38 185ML
SDP
Catalog No. 501861
Encompass
HR2-922-38/PEG/Ion Screen #38 (D2), 185 ml - 0.2 M Ammonium tartrate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-1 Supplier Hampton Research Supplier No. HR292238
HR2-922-38/PEG/Ion Screen #38 (D2), 185 ml - 0.2 M Ammonium tartrate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.