Learn More
Hampton Research PEG/ION SCREEN NO39 185ML
SDP
Catalog No. 501862
Encompass
HR2-922-39/PEG/Ion Screen #39 (D3), 185 ml - 0.2 M Sodium phosphate monobasic monohydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-2 Supplier Hampton Research Supplier No. HR292239
HR2-922-39/PEG/Ion Screen #39 (D3), 185 ml - 0.2 M Sodium phosphate monobasic monohydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.