Learn More
Hampton Research PEG/ION SCREEN NO40 185ML
SDP
Catalog No. 501863
Encompass
HR2-922-40/PEG/Ion Screen #40 (D4), 185 ml - 0.2 M Sodium phosphate dibasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-3 Supplier Hampton Research Supplier No. HR292240
HR2-922-40/PEG/Ion Screen #40 (D4), 185 ml - 0.2 M Sodium phosphate dibasic dihydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.