Learn More
Hampton Research PEG/ION SCREEN NO42 185ML
SDP
Catalog No. 501865
Encompass
HR2-922-42/PEG/Ion Screen #42 (D6), 185 ml - 0.2 M Potassium phosphate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-5 Supplier Hampton Research Supplier No. HR292242
HR2-922-42/PEG/Ion Screen #42 (D6), 185 ml - 0.2 M Potassium phosphate dibasic, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.