Learn More
Hampton Research PEG/ION SCREEN NO45 185ML
SDP
Catalog No. 501868
Encompass
HR2-922-45/PEG/Ion Screen #45 (D9), 185 ml - 0.2 M Lithium citrate tribasic tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-186-8 Supplier Hampton Research Supplier No. HR292245
HR2-922-45/PEG/Ion Screen #45 (D9), 185 ml - 0.2 M Lithium citrate tribasic tetrahydrate, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.